accessibility menu, dialog, popup

UserName

CAS RN 61-82-5

IUPAC Name: 1H-1,2,4-triazol-5-amine
Synonyms: 5-amino-1H-1,2,4-triazole amitrole (iso)
Molecular Weight (g/mol): 84.08
Molecular Formula: C2H4N4
InChi Key: KLSJWNVTNUYHDU-UHFFFAOYSA-N
SMILES: NC1=NC=NN1
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)