missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-(tert-butoxycarbonyl)pyrrolidine 98.0+%, TCI America™

Catalog No. B38901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38901G 1 g
B38905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38901G Supplier TCI America Supplier No. B38901G

Chemical Identifiers

CAS 186550-13-0
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD01861220
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-O
Synonym 1-boc-3-aminopyrrolidine, 3-amino-1-boc-pyrrolidine, +/--3-amino-1-n-boc-pyrrolidine, 3-amino-1-n-boc-pyrrolidine, 3-amino-1-tert-butoxycarbonyl pyrrolidine, +/--1-boc-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, n-boc-3-aminopyrrolidine, 1-bocap
PubChem CID 2756370
IUPAC Name (3R)-1-[(tert-butoxy)carbonyl]pyrrolidin-3-aminium
SMILES CC(C)(C)OC(=O)N1CC[C@@H]([NH3+])C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-(tert-butoxycarbonyl)pyrrolidine
TSCA No
Recommended Storage Refrigerator