accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(R)H3N

CAS RN 186550-13-0

IUPAC Name: (3R)-1-[(tert-butoxy)carbonyl]pyrrolidin-3-aminium
Synonyms: (3R)-1-(tert-butoxycarbonyl)pyrrolidin-3-aminium
Molecular Weight (g/mol): 187.26
Molecular Formula: C9H19N2O2
InChi Key: CMIBWIAICVBURI-SSDOTTSWSA-O
SMILES: CC(C)(C)OC(=O)N1CC[C@@H]([NH3+])C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)