missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-propanol Hydrochloride 95.0+%, TCI America™

Catalog No. a040725g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A040725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A040725G Supplier TCI America Supplier No. A040725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14302-46-6
Molecular Formula C3H10ClNO
Molecular Weight (g/mol) 111.569
MDL Number MFCD00060203
InChI Key RIAJTMUAPNIDSS-UHFFFAOYSA-N
Synonym Propanolamine Hydrochloride
PubChem CID 3014826
IUPAC Name 3-aminopropan-1-ol;hydrochloride
SMILES C(CN)CO.Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 111.57
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-propanol Hydrochloride
EINECSNumber (2)-0323
TSCA No