accessibility menu, dialog, popup

UserName

HClNH2HO

CAS RN 14302-46-6

IUPAC Name: hydrogen 3-aminopropan-1-ol chloride
Synonyms: hydrogen propanolamine chloride
Molecular Weight (g/mol): 111.57
Molecular Formula: C3H10ClNO
InChi Key: RIAJTMUAPNIDSS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results