missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methylpyrazole 98.0+%, TCI America™

Catalog No. p-7117819
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22771G 1 g
A22775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22771G Supplier TCI America Supplier No. A22771G
Only null left

Chemical Identifiers

CAS 1904-31-0
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00466340
InChI Key MOGQNVSKBCVIPW-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole
PubChem CID 137254
IUPAC Name 1-methylpyrazol-3-amine
SMILES CN1C=CC(=N1)N

Specifications

Color Yellow
Boiling Point 89°C
Quantity 1 g
Formula Weight 97.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-1-methylpyrazole