missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methyl-1H-pyrazole, 97%

Catalog No. AAH3093503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3093503 1 g
AAH3093506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3093503 Supplier Thermo Scientific Chemicals Supplier No. H3093503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1904-31-0 | C4H7N3 | 97.121 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1904-31-0
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00466340
InChI Key MOGQNVSKBCVIPW-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole
PubChem CID 137254
IUPAC Name 1-methylpyrazol-3-amine
SMILES CN1C=CC(=N1)N

Specifications

Boiling Point 90°C to 92°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 3-Amino-1-methyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only