missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-Boc-piperidine, 97%

Catalog No. p-7035109
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281603 1 g
AAH5281606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5281603 Supplier Thermo Scientific Chemicals Supplier No. H5281603
Only null left

CAS: 184637-48-7 | C10H20N2O2 | 200.282 g/mol

3-Amino-1-Boc-piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-Boc-piperidine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 184637-48-7
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD01861219
InChI Key AKQXKEBCONUWCL-UHFFFAOYSA-N
Synonym 1-boc-3-aminopiperidine, n-boc-3-aminopiperidine, 3-amino-1-n-boc-piperidine, +/--3-amino-1-n-boc-piperidine, 3-amino-1-boc-piperidine, 1-n-boc-3-aminopiperidine, 3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3-amino-1-tert-butoxycarbonylpiperidine, 3-aminopiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 545809
IUPAC Name tert-butyl 3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)N

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 3-Amino-1-Boc-piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only