accessibility menu, dialog, popup

UserName

CAS RN 184637-48-7

IUPAC Name: hydrogen tert-butyl 3-aminopiperidine-1-carboxylate chloride
Synonyms: hydrogen tert-butyl 3-aminopiperidine-1-carboxylate chloride
Molecular Weight (g/mol): 236.74
Molecular Formula: C10H21ClN2O2
InChi Key: DUNAVVRRZJQGCZ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(C)OC(=O)N1CCCC(N)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results