missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1-benzylpiperidine 98.0+%, TCI America™
$154.71 - $462.29
Chemical Identifiers
| CAS | 60407-35-4 |
|---|---|
| Molecular Formula | C12H18N2 |
| Molecular Weight (g/mol) | 190.29 |
| MDL Number | MFCD04038444 |
| InChI Key | HARWNWOLWMTQCC-UHFFFAOYSA-N |
| PubChem CID | 421694 |
| IUPAC Name | 1-benzylpiperidin-3-amine |
| SMILES | C1CC(CN(C1)CC2=CC=CC=C2)N |
Chemical Identifiers
| 60407-35-4 | |
| 190.29 | |
| HARWNWOLWMTQCC-UHFFFAOYSA-N | |
| 1-benzylpiperidin-3-amine |
| C12H18N2 | |
| MFCD04038444 | |
| 421694 | |
| C1CC(CN(C1)CC2=CC=CC=C2)N |
Specifications
| 60407-35-4 | |
| 82°C | |
| MFCD04038444 | |
| HARWNWOLWMTQCC-UHFFFAOYSA-N | |
| 1-benzylpiperidin-3-amine | |
| 421694 | |
| ≥98.0% (GC,T) | |
| 3-Amino-1-benzylpiperidine |
| White-Yellow | |
| C12H18N2 | |
| 1 g | |
| C1CC(CN(C1)CC2=CC=CC=C2)N | |
| 190.29 | |
| 190.29 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator