missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-(4-chlorophenyl)-1H-pyrazole, 95%, Thermo Scientific™

Catalog No. AAH32252MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32252MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32252MD Supplier Thermo Scientific Chemicals Supplier No. H32252MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 66000-39-3
Molecular Formula C9H8ClN3
Molecular Weight (g/mol) 193.634
MDL Number MFCD09027932
InChI Key SHCVEWFQFUKSDP-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-pyrazol-3-amine, 3-amino-1-4-chlorophenyl-1h-pyrazole, 1-4-chlorophenyl pyrazol-3-amine, 1-p-chlorophenyl-3-amino-pyrazole, 1-4-chlorophenyl-1h-pyrazol-3-ylamine, 1-4-chloro-phenyl-1h-pyrazol-3-ylamine
PubChem CID 13626945
IUPAC Name 1-(4-chlorophenyl)pyrazol-3-amine
SMILES C1=CC(=CC=C1N2C=CC(=N2)N)Cl

Specifications

Melting Point 107°C to 108°C
Quantity 250 mg
Formula Weight 193.64
Percent Purity 95%
Chemical Name or Material 3-Amino-1-(4-chlorophenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures