accessibility menu, dialog, popup

UserName

CAS RN 66000-39-3

IUPAC Name: 1-(4-chlorophenyl)-1H-pyrazol-3-amine
Synonyms: 1-(4-chlorophenyl)pyrazol-3-amine
Molecular Weight (g/mol): 193.63
Molecular Formula: C9H8ClN3
InChi Key: SHCVEWFQFUKSDP-UHFFFAOYSA-N
SMILES: NC1=NN(C=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results