missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazole-5-carboxylic Acid Hemihydrate 98.0+%, TCI America™

Catalog No. a125625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A125625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A125625G Supplier TCI America Supplier No. A125625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3641-13-2
Molecular Formula C3H4N4O2
Molecular Weight (g/mol) 128.09
MDL Number MFCD00010620 MFCD00179325 MFCD00225624
InChI Key MVRGLMCHDCMPKD-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole-5-carboxylic acid, 5-amino-4h-1,2,4-triazole-3-carboxylic acid, 5-amino-1h-1,2,4-triazole-3-carboxylic acid, 1h-1,2,4-triazole-3-carboxylic acid, 5-amino, 5-amino-1,2,4-triazole-3-carboxylic acid, s-triazole-3-carboxylic acid, 5-amino, 5-amino-2h-1,2,4-triazole-3-carboxylic acid, 5-amino-1h-1,2,4 triazole-3-carboxylic acid, 5-amino-2h-1,2,4 triazole-3-carboxylic acid, 1h-1,4-triazole-3-carboxylic acid, 5-amino
PubChem CID 77200
IUPAC Name 3-amino-1H-1,2,4-triazole-5-carboxylic acid
SMILES NC1=NNC(=N1)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 128.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1,2,4-triazole-5-carboxylic Acid Hemihydrate