accessibility menu, dialog, popup

UserName

CAS RN 3641-13-2

NH2NNHNOHO

CAS RN 3641-13-2

IUPAC Name: 3-amino-1H-1,2,4-triazole-5-carboxylic acid
Synonyms: 5-amino-2H-1,2,4-triazole-3-carboxylic acid
Molecular Weight (g/mol): 128.09
Molecular Formula: C3H4N4O2
InChi Key: MVRGLMCHDCMPKD-UHFFFAOYSA-N
SMILES: NC1=NNC(=N1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results