missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acryloyl-2-oxazolidinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 141.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Acryloyl-2-oxazolidinone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |