accessibility menu, dialog, popup

UserName

CAS RN 2043-21-2

CH2ONOO

CAS RN 2043-21-2

IUPAC Name: 3-(prop-2-enoyl)-1,3-oxazolidin-2-one
Synonyms: 3-(prop-2-enoyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: HIBSYUPTCGGRSD-UHFFFAOYSA-N
SMILES: C=CC(=O)N1CCOC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results