missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine 98.0+%, TCI America™

Catalog No. A011225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A011225G 25 g
A0112250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A011225G Supplier TCI America Supplier No. A011225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
UN Number 2810
Formula Weight 121.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetylpyridine
EINECSNumber (5)-3706
RTECSNumber OB5425000
TSCA Yes