accessibility menu, dialog, popup

UserName

CAS RN 350-03-8

IUPAC Name: 1-(pyridin-3-yl)ethan-1-one
Synonyms: 3-acetylpyridine
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: WEGYGNROSJDEIW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CN=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)