missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine, 98%

CAS: 350-03-8 | C7H7NO | 121.14 g/mol

$58.61 - $997.64

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102690100
SDS View Documents
Thermo Scientific Chemicals
102690100
10 g Glass bottle
Each for $58.61
Only null left
 
AC102691000
SDS View Documents
Thermo Scientific Chemicals
102691000
100 g Glass bottle
Each for $308.98
Only null left
 
AC102695000
SDS View Documents
Thermo Scientific Chemicals
102695000
500 g Glass bottle
Each for $997.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

350-03-8
121.14
WEGYGNROSJDEIW-UHFFFAOYSA-N
9589
CC(=O)C1=CN=CC=C1
C7H7NO
MFCD00006396
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
1-pyridin-3-ylethanone

Specifications

350-03-8
100.0
Colorless to Yellow
220.0°C
Authentic
Glass bottle
1.5320 to 1.5350
MFCD00006396
21,279
15,6189
Solubility in water: soluble in hot water. Other solubilities: soluble in acids,miscible with organic solvents except ligroin
CC(=O)C1=CN=CC=C1
121.14
121.14
Liquid
97.5
12.0°C to 13.0°C
1.1000g/mL
104°C
97.5% min. (GC)
C7H7NO
(C5H4N)COCH3
10 g
1.1
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
WEGYGNROSJDEIW-UHFFFAOYSA-N
1-pyridin-3-ylethanone
9589
98%
3-Acetylpyridine, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou

GHS Signal Word: Danger

EINECSNumber : 206-496-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only