missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylpyridine, 98%

Catalog No. AC102691000
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102691000 100 g Glass bottle
AC102690100 10 g Glass bottle
AC102695000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102691000 Supplier Thermo Scientific Chemicals Supplier No. 102691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 350-03-8 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 12.0°C to 13.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 220.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5350
Linear Formula (C5H4N)COCH3
Quantity 100 g
Beilstein 21,279
Merck Index 15,6189
Specific Gravity 1.1
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in acids,miscible with organic solvents except ligroin
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Acetylpyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou
EINECSNumber 206-496-7

RUO – Research Use Only