missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylpyridine, 98%
Description
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
Solubility
Soluble in Chloroform. Miscible with water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
Solubility
Soluble in Chloroform. Miscible with water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 12°C to 13°C |
| Density | 1.106 |
| Boiling Point | 218°C to 220°C |
| Flash Point | 104°C (219°F) |
| Odor | Characteristic |
| Refractive Index | 1.535 |
| Quantity | 25 g |
| UN Number | UN2810 |
| Beilstein | 107751 |
| Merck Index | 14,6116 |
| Show More |
Safety and Handling
| Hazard Category | H300-H315-H319 |
| Hazard Statement | GHS H Statement H301-H315-H319 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 206-496-7 |
| RTECSNumber | OB5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only