missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetyldeoxynivalenol, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 50722-38-8 |
| Physical Form | Neat |
| Linear Formula | C17H22O7 |
| MDL Number | MFCD00135964 |
| Synonym | 3 alpha-Acetoxy-7 alpha,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one; 3 alpha-Acetylvomitoxin; 3-AcDON |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 338.35 |
| Grade | Analytical Standard |