missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylcoumarin, 98+%

Catalog No. aaa1253214
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAA1253214 25 g
AAA1253206 5 g
2 options

Catalog No. AAA1253214

Supplier: Thermo Scientific Chemicals A1253214

Only null left

CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3949-36-8
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00006853
InChI Key CSPIFKKOBWYOEX-UHFFFAOYSA-N
Synonym 3-acetylcoumarin, 3-acetyl-2h-chromen-2-one, coumarin, 3-acetyl, 2h-1-benzopyran-2-one, 3-acetyl, 3-acetyl-2-oxochromene, 3-acetylcoumarine, pubchem8664, maybridge1_000885, prestwick0_000891, prestwick1_000891
PubChem CID 77553
IUPAC Name 3-acetylchromen-2-one
SMILES CC(=O)C1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 119°C to 124°C
Quantity 25 g
Beilstein 143498
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material 3-Acetylcoumarin
EINECSNumber 223-541-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only