accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 3949-36-8

IUPAC Name: 3-acetyl-2H-chromen-2-one
Synonyms: 3-acetylchromen-2-one
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: CSPIFKKOBWYOEX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC=CC=C2OC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)