Learn More
3-Acetylbenzonitrile, 97+%
CAS: 6136-68-1 | C9H7NO | 145.16 g/mol
$130.47 - $453.68
Chemical Identifiers
| CAS | 6136-68-1 |
|---|---|
| Molecular Formula | C9H7NO |
| Molecular Weight (g/mol) | 145.16 |
| MDL Number | MFCD00001806 |
| InChI Key | SBCFGFDAZCTSRH-UHFFFAOYSA-N |
| Synonym | m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 |
| PubChem CID | 80222 |
| IUPAC Name | 3-acetylbenzonitrile |
| SMILES | CC(=O)C1=CC=CC(=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1084106
|
Thermo Scientific Chemicals
A1084106 |
5 g |
Each for $130.47
|
|
|||||
|
AAA1084114
|
Thermo Scientific Chemicals
A1084114 |
25 g |
Each for $453.68
|
|
|||||
Description
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPreparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 6136-68-1 | |
| 145.16 | |
| SBCFGFDAZCTSRH-UHFFFAOYSA-N | |
| 80222 | |
| CC(=O)C1=CC=CC(=C1)C#N |
| C9H7NO | |
| MFCD00001806 | |
| m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 | |
| 3-acetylbenzonitrile |
Specifications
| 6136-68-1 | |
| 120°C (5 mmHg) | |
| MFCD00001806 | |
| UN3439 | |
| m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 | |
| SBCFGFDAZCTSRH-UHFFFAOYSA-N | |
| 3-acetylbenzonitrile | |
| 80222 | |
| ≥97% |
| 97°C to 101°C | |
| C9H7NO | |
| 5 g | |
| 2079629 | |
| Insoluble in water. | |
| CC(=O)C1=CC=CC(=C1)C#N | |
| 145.16 | |
| 145.16 | |
| 3-Acetylbenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 228-110-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only