missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylbenzoic Acid 97.0+%, TCI America™
$438.05 - $1757.81
Chemical Identifiers
| CAS | 586-42-5 |
|---|---|
| Molecular Formula | C9H8O3 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00045847 |
| InChI Key | CHZPJUSFUDUEMZ-UHFFFAOYSA-N |
| Synonym | m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid |
| PubChem CID | 220324 |
| IUPAC Name | 3-acetylbenzoic acid |
| SMILES | CC(=O)C1=CC(=CC=C1)C(=O)O |
Chemical Identifiers
| 586-42-5 | |
| 164.16 | |
| CHZPJUSFUDUEMZ-UHFFFAOYSA-N | |
| 220324 | |
| CC(=O)C1=CC(=CC=C1)C(=O)O |
| C9H8O3 | |
| MFCD00045847 | |
| m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid | |
| 3-acetylbenzoic acid |
Specifications
| 586-42-5 | |
| Yellow | |
| MFCD00045847 | |
| m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid | |
| CC(=O)C1=CC(=CC=C1)C(=O)O | |
| 164.16 | |
| 164.16 | |
| Crystalline Powder |
| 164°C | |
| C9H8O3 | |
| 1 g | |
| CHZPJUSFUDUEMZ-UHFFFAOYSA-N | |
| 3-acetylbenzoic acid | |
| 220324 | |
| ≥97.0% (GC,T) | |
| 3-Acetylbenzoic Acid |
Safety and Handling
TSCA : No