accessibility menu, dialog, popup

UserName

CAS RN 586-42-5

IUPAC Name: 3-acetylbenzoate
Synonyms: 3-acetylbenzoate
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H7O3
InChi Key: CHZPJUSFUDUEMZ-UHFFFAOYSA-M
SMILES: CC(=O)C1=CC=CC(=C1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)