missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzoic Acid 97.0+%, TCI America™

Catalog No. A16525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16525G 5 g
A16521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16525G Supplier TCI America Supplier No. A16525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 586-42-5
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00045847
InChI Key CHZPJUSFUDUEMZ-UHFFFAOYSA-N
Synonym m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid
PubChem CID 220324
IUPAC Name 3-acetylbenzoic acid
SMILES CC(=O)C1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 164°C
Color Yellow
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetylbenzoic Acid