Learn More
3-Acetylbenzoic acid, 98%
CAS: 586-42-5 | C9H8O3 | 164.16 g/mol
$190.66 - $582.55
Chemical Identifiers
| CAS | 586-42-5 |
|---|---|
| Molecular Formula | C9H8O3 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00045847 |
| InChI Key | CHZPJUSFUDUEMZ-UHFFFAOYSA-N |
| Synonym | m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid |
| PubChem CID | 220324 |
| IUPAC Name | 3-acetylbenzoic acid |
| SMILES | CC(=O)C1=CC(=CC=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2561603
|
Thermo Scientific Chemicals
B2561603 |
1 g |
N/A
|
|
|||||
|
AAB2561606
|
Thermo Scientific Chemicals
B2561606 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 586-42-5 | |
| 164.16 | |
| CHZPJUSFUDUEMZ-UHFFFAOYSA-N | |
| 220324 | |
| CC(=O)C1=CC(=CC=C1)C(=O)O |
| C9H8O3 | |
| MFCD00045847 | |
| m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid | |
| 3-acetylbenzoic acid |
Specifications
| 586-42-5 | |
| Odorless | |
| MFCD00045847 | |
| m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid | |
| CC(=O)C1=CC(=CC=C1)C(=O)O | |
| 164.16 | |
| 164.16 | |
| 3-Acetylbenzoic acid |
| 162°C to 166°C | |
| C9H8O3 | |
| 1 g | |
| CHZPJUSFUDUEMZ-UHFFFAOYSA-N | |
| 3-acetylbenzoic acid | |
| 220324 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only