missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzoic acid, 98%

Catalog No. AAB2561603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2561603 1 g
AAB2561606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2561603 Supplier Thermo Scientific Chemicals Supplier No. B2561603
Only null left

CAS: 586-42-5 | C9H8O3 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 586-42-5
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00045847
InChI Key CHZPJUSFUDUEMZ-UHFFFAOYSA-N
Synonym m-acetylbenzoic acid, benzoic acid, 3-acetyl, 3-acetylbenzoicacid, 3-acetophenonecarboxylic acid, acetophenone-3-carboxylic acid, 3-acetyl benzoic acid, 3-acetyl-benzoic acid, hypnone-m-carboxylic acid, 3-1-oxoethyl benzoic acid, 3-acetylbenzoic acid
PubChem CID 220324
IUPAC Name 3-acetylbenzoic acid
SMILES CC(=O)C1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 162°C to 166°C
Odor Odorless
Quantity 1 g
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 3-Acetylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only