missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylbenzenesulfonyl chloride, 97%

Catalog No. AAH3244703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3244703 1 g
AAH3244706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3244703 Supplier Thermo Scientific Chemicals Supplier No. H3244703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73035-16-2 | C8H7ClO3S | 218.651 g/mol

Classical acetamidomalonate addition/decarboxylative hydrolysis as the key step for the generation of the amino acidic moiety starting from 4-ethylsulfonyl-α-bromoacetophenone prepared in two steps from commercially available 4-acetyl-benzenesulfonyl chloride. This then treated with sodium sulfite in basic aqueous conditions yielded the sodium sulfinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Classical acetamidomalonate addition/decarboxylative hydrolysis as the key step for the generation of the amino acidic moiety starting from 4-ethylsulfonyl-α-bromoacetophenone prepared in two steps from commercially available 4-acetyl-benzenesulfonyl chloride. This then treated with sodium sulfite in basic aqueous conditions yielded the sodium sulfinate.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

Chemical Identifiers

CAS 73035-16-2
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.651
MDL Number MFCD03424979
InChI Key CGMBNEIGZOCPPP-UHFFFAOYSA-N
Synonym 3-acetylbenzene-1-sulfonyl chloride, 3-acetylphenylsulfonyl chloride, 3-acetyl benzenesulfonyl chloride, benzenesulfonylchloride, 3-acetyl, m-chlorosulfonylacetophenone, 3-acetyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-acetyl, benzene sulfonylchloride 3-acetyl, benzenesulfonyl chloride, 3-acetyl-9ci
PubChem CID 2756260
IUPAC Name 3-acetylbenzenesulfonyl chloride
SMILES CC(=O)C1=CC(=CC=C1)S(=O)(=O)Cl

Specifications

Melting Point 42°C to 43°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 218.66
Percent Purity 97%
Chemical Name or Material 3-Acetylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only