CAS RN 73035-16-2
CAS RN 73035-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Acetylbenzenesulfonyl chloride, 97%
CAS: 73035-16-2 Molecular Formula: C8H7ClO3S Molecular Weight (g/mol): 218.651 MDL Number: MFCD03424979 InChI Key: CGMBNEIGZOCPPP-UHFFFAOYSA-N Synonym: 3-acetylbenzene-1-sulfonyl chloride,3-acetylphenylsulfonyl chloride,3-acetyl benzenesulfonyl chloride,benzenesulfonylchloride, 3-acetyl,m-chlorosulfonylacetophenone,3-acetyl-benzenesulfonyl chloride,benzenesulfonyl chloride, 3-acetyl,benzene sulfonylchloride 3-acetyl,benzenesulfonyl chloride, 3-acetyl-9ci PubChem CID: 2756260 IUPAC Name: 3-acetylbenzenesulfonyl chloride SMILES: CC(=O)C1=CC(=CC=C1)S(=O)(=O)Cl