missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-5-bromopyridine 97.0+%, TCI America™

Catalog No. A23761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23761G 1 g
A23765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23761G Supplier TCI America Supplier No. A23761G

Chemical Identifiers

CAS 38940-62-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.04
MDL Number MFCD03086033
InChI Key LDBPZEQZCOUYFT-UHFFFAOYSA-N
Synonym 3-acetyl-5-bromopyridine, 1-5-bromopyridin-3-yl ethanone, 1-5-bromopyridin-3-yl ethan-1-one, ethanone, 1-5-bromo-3-pyridinyl, 3-bromo-5-acetylpyridine, 1-5-bromo-3-pyridyl ethanone, 5-acetyl-3-bromopyridine, 1-5-bromo-3-pyridinyl ethanone, pubchem15626, acmc-1aglt
PubChem CID 820423
IUPAC Name 1-(5-bromopyridin-3-yl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=CN=C1

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-5-bromopyridine