accessibility menu, dialog, popup

UserName

H3CONBr

CAS RN 38940-62-4

IUPAC Name: 1-(5-bromopyridin-3-yl)ethan-1-one
Synonyms: 1-(5-bromopyridin-3-yl)ethanone
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: LDBPZEQZCOUYFT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=CN=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)