missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-2,5-dichlorothiophene 98.0+%, TCI America™

Catalog No. A11825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11825G 5 g
A118225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11825G Supplier TCI America Supplier No. A11825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36157-40-1
Molecular Formula C6H4Cl2OS
Molecular Weight (g/mol) 195.057
MDL Number MFCD00014522
InChI Key GYFDNIRENHZKGR-UHFFFAOYSA-N
Synonym 3-acetyl-2,5-dichlorothiophene, 1-2,5-dichlorothiophen-3-yl ethanone, 1-2,5-dichloro-3-thienyl ethan-1-one, 1-2,5-dichlorothiophen-3-yl ethan-1-one, 1-2,5-dichloro-3-thienyl ethanone, 2,5-dichloro-3-thienyl methyl ketone, ethanone, 1-2,5-dichloro-3-thienyl, 1-2,5-dichloro-thiophen-3-yl-ethanone, 1-2,5-dichlorothien-3-yl ethan-1-one, zlchem 481
PubChem CID 118920
IUPAC Name 1-(2,5-dichlorothiophen-3-yl)ethanone
SMILES CC(=O)C1=C(SC(=C1)Cl)Cl

Specifications

Melting Point 38°C
Boiling Point 122°C
Quantity 5 g
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-2,5-dichlorothiophene