accessibility menu, dialog, popup

UserName

H3COClSCl

CAS RN 36157-40-1

IUPAC Name: 1-(2,5-dichlorothiophen-3-yl)ethan-1-one
Synonyms: 1-(2,5-dichlorothiophen-3-yl)ethanone
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H4Cl2OS
InChi Key: GYFDNIRENHZKGR-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)SC(Cl)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)