missing translation for 'onlineSavingsMsg'
Learn More

3'-Acetoxyacetophenone 98.0+%, TCI America™

Catalog No. A19965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19965G 5 g
A199625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19965G Supplier TCI America Supplier No. A19965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2454-35-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00017227
InChI Key OTHYPAMNTUGKDK-UHFFFAOYSA-N
PubChem CID 75563
IUPAC Name (3-acetylphenyl) acetate
SMILES CC(=O)C1=CC(=CC=C1)OC(=O)C

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 147°C
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Acetoxyacetophenone