accessibility menu, dialog, popup

UserName

CH3OOCH3O

CAS RN 2454-35-5

IUPAC Name: 3-acetylphenyl acetate
Synonyms: 3-acetylphenyl acetate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: OTHYPAMNTUGKDK-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results