missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-2-naphthanilide 99.0+%, TCI America™

Catalog No. A00695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A00695G Supplier TCI America Supplier No. A00695G

Chemical Identifiers

CAS 1163-67-3
Molecular Formula C19H15NO3
Molecular Weight (g/mol) 305.333
MDL Number MFCD00004095
InChI Key CVJGNNYDVQYHEO-UHFFFAOYSA-N
Synonym naphthol as acetate, 3-acetoxy-2-naphthanilide, 2-acetoxy-3-naphthoic acid anilide, acetyl naphthol as, 3-phenylcarbamoyl naphthalen-2-yl acetate, 2-n-phenylcarbamoyl-3-naphthyl acetate, 2-naphthalenecarboxamide, 3-acetyloxy-n-phenyl, naphthol-as-acetat, 3-phenylcarbamoyl-2-naphthyl acetate
PubChem CID 96045
IUPAC Name [3-(phenylcarbamoyl)naphthalen-2-yl] acetate
SMILES CC(=O)OC1=CC2=CC=CC=C2C=C1C(=O)NC3=CC=CC=C3

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 305.33
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetoxy-2-naphthanilide