missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetoxy-2-methylbenzoic Acid 98.0+%, TCI America™
$59.65 - $188.27
Chemical Identifiers
| CAS | 168899-58-9 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.186 |
| MDL Number | MFCD00957176 |
| InChI Key | LHVJUPHCLWIPLZ-UHFFFAOYSA-N |
| Synonym | 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid |
| PubChem CID | 3798557 |
| IUPAC Name | 3-acetyloxy-2-methylbenzoic acid |
| SMILES | CC1=C(C=CC=C1OC(=O)C)C(=O)O |
Chemical Identifiers
| 168899-58-9 | |
| 194.186 | |
| LHVJUPHCLWIPLZ-UHFFFAOYSA-N | |
| 3798557 | |
| CC1=C(C=CC=C1OC(=O)C)C(=O)O |
| C10H10O4 | |
| MFCD00957176 | |
| 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid | |
| 3-acetyloxy-2-methylbenzoic acid |
Specifications
| 168899-58-9 | |
| White | |
| MFCD00957176 | |
| 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid | |
| CC1=C(C=CC=C1OC(=O)C)C(=O)O | |
| 194.186 | |
| 194.19 | |
| Crystalline Powder |
| 148°C | |
| C10H10O4 | |
| 5 g | |
| LHVJUPHCLWIPLZ-UHFFFAOYSA-N | |
| 3-acetyloxy-2-methylbenzoic acid | |
| 3798557 | |
| ≥98.0% (HPLC,T) | |
| 3-Acetoxy-2-methylbenzoic Acid |
Safety and Handling
EINECSNumber : (3)-4420&(3)-4455
TSCA : No