missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A24531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24531G Supplier TCI America Supplier No. A24531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Acetamidophenylboronic Acid (contains varying amounts of Anhydride)
CAS 78887-39-5
Melting Point 140°C
Color Yellow
Molecular Formula C8H10BNO3
Quantity 1 g
MDL Number MFCD00236013
Synonym 3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid
InChI Key IBTSWKLSEOGJGJ-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)C)(O)O
IUPAC Name (3-acetamidophenyl)boronic acid
Molecular Weight (g/mol) 178.982
PubChem CID 157274
Formula Weight 178.98
Physical Form Crystalline Powder
Show More Show Less