missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidophenol, 98%

Catalog No. AAA1276414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276414 25 g
AAA1276422 100 g
AAA1276436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1276414 Supplier Thermo Scientific Chemicals Supplier No. A1276414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-42-1 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-42-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002263
InChI Key QLNWXBAGRTUKKI-UHFFFAOYSA-N
Synonym 3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
PubChem CID 12124
ChEBI CHEBI:76987
IUPAC Name N-(3-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)O

Specifications

Melting Point 146°C to 149°C
Quantity 25 g
Beilstein 907998
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 3-Acetamidophenol
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-687-0
RTECSNumber AE4100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only