missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidobenzoic Acid 99.0+%, TCI America™

Catalog No. A001225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A001225G Supplier TCI America Supplier No. A001225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 587-48-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00013983
InChI Key RGDPZMQZWZMONQ-UHFFFAOYSA-N
Synonym 3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
PubChem CID 48847
IUPAC Name 3-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 248°C
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetamidobenzoic Acid
EINECSNumber (3)-1450
TSCA Yes