missing translation for 'onlineSavingsMsg'
Learn More

3-Acetamidobenzoic acid, 98%

Catalog No. AAA1590222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590222 100 g
AAA1590214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1590222 Supplier Thermo Scientific Chemicals Supplier No. A1590222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 587-48-4 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 587-48-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00013983
InChI Key RGDPZMQZWZMONQ-UHFFFAOYSA-N
Synonym 3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
PubChem CID 48847
IUPAC Name 3-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 249°C to 252°C
Quantity 100 g
Beilstein 2210867
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 3-Acetamidobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-600-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only