missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(9H-Carbazol-9-yl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$68.97 - $68.97
Specifications
| CAS | 864377-33-3 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C18H14BNO2 |
| MDL Number | MFCD18072506 |
| InChI Key | IDQUIFLAFFZYEX-UHFFFAOYSA-N |
Specifications
| 864377-33-3 | |
| C18H14BNO2 | |
| IDQUIFLAFFZYEX-UHFFFAOYSA-N | |
| (3-carbazol-9-ylphenyl)boronic acid | |
| 287.13 |
| White-Yellow | |
| MFCD18072506 | |
| B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O | |
| 287.125 |