accessibility menu, dialog, popup

UserName

BNOHOH

CAS RN 864377-33-3

IUPAC Name: [3-(9H-carbazol-9-yl)phenyl]boronic acid
Synonyms: 3-(carbazol-9-yl)phenylboronic acid
Molecular Weight (g/mol): 287.13
Molecular Formula: C18H14BNO2
InChi Key: IDQUIFLAFFZYEX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)