missing translation for 'onlineSavingsMsg'
Learn More

3-(9-Carbazolyl)benzeneboronic acid, 98%

CAS: 864377-33-3 | C18H14BNO2 | 287.125 g/mol

$95.85 - $307.16

Chemical Identifiers

CAS 864377-33-3
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.125
MDL Number MFCD18072506
InChI Key IDQUIFLAFFZYEX-UHFFFAOYSA-N
Synonym 3-9h-carbazol-9-yl phenylboronic acid, 3-9h-carbazol-9-yl phenyl boronic acid, boronic acid, 3-9h-carbazol-9-yl phenyl, 3-9-carbazolyl benzeneboronic acid, 3-carbazol-9-yl phenylboronic acid, 3-9h-9-carbozale phenylboronic acid, 3-9h-9-carbazole phenylboronic acid, 3-9h-carbazol-9-yl benzeneboronic acid
PubChem CID 56965778
IUPAC Name (3-carbazol-9-ylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6489803
View Documents
Thermo Scientific Chemicals
H6489803
1 g
Each for $95.85
Only null left
 
AAH6489806
View Documents
Thermo Scientific Chemicals
H6489806
5 g
Each for $307.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

864377-33-3
287.125
IDQUIFLAFFZYEX-UHFFFAOYSA-N
56965778
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
C18H14BNO2
MFCD18072506
3-9h-carbazol-9-yl phenylboronic acid, 3-9h-carbazol-9-yl phenyl boronic acid, boronic acid, 3-9h-carbazol-9-yl phenyl, 3-9-carbazolyl benzeneboronic acid, 3-carbazol-9-yl phenylboronic acid, 3-9h-9-carbozale phenylboronic acid, 3-9h-9-carbazole phenylboronic acid, 3-9h-carbazol-9-yl benzeneboronic acid
(3-carbazol-9-ylphenyl)boronic acid

Specifications

864377-33-3
C18H14BNO2
1 g
3-9h-carbazol-9-yl phenylboronic acid, 3-9h-carbazol-9-yl phenyl boronic acid, boronic acid, 3-9h-carbazol-9-yl phenyl, 3-9-carbazolyl benzeneboronic acid, 3-carbazol-9-yl phenylboronic acid, 3-9h-9-carbozale phenylboronic acid, 3-9h-9-carbazole phenylboronic acid, 3-9h-carbazol-9-yl benzeneboronic acid
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
287.125
287.13
Powder
White
MFCD18072506
15530380
IDQUIFLAFFZYEX-UHFFFAOYSA-N
(3-carbazol-9-ylphenyl)boronic acid
56965778
98%
3-(9-Carbazolyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only