missing translation for 'onlineSavingsMsg'
Learn More

3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane 98.0+%, TCI America™

Catalog No. p-7118885
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B115825G 25 g
B1158500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B115825G Supplier TCI America Supplier No. B115825G
Only null left

Chemical Identifiers

CAS 1455-42-1
Molecular Formula C15H28O6
Molecular Weight (g/mol) 304.383
MDL Number MFCD00059794
InChI Key BPZIYBJCZRUDEG-UHFFFAOYSA-N
Synonym 2,2'-2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diyl bis 2-methylpropan-1-ol, 3,9-bis 1,1-dimethyl-2-hydroxyethyl-2,4,8,10-tetraoxaspiro 5.5 undecane, cyclic acetal, 2-9-1-hydroxy-2-methylpropan-2-yl-2,4,8,10-tetraoxaspiro 5.5 undecan-3-yl-2-methylpropan-1-ol, acmc-209cve, ksc489m7h, 2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diethanol, beta3,beta3,beta9,beta9-tetramethyl, bis hydroxypivalaldehyde pentaerythritolacetal, bis hydroxypivalaldehyde pentaerythritolacetal cyclic acetal, bis hydroxypivalaldehyde pentaerythritol acetal cyclic acetal
PubChem CID 7569008
IUPAC Name 2-[3-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]-2-methylpropan-1-ol
SMILES CC(C)(CO)C1OCC2(CO1)COC(OC2)C(C)(C)CO

Specifications

Melting Point 196°C
Color White
Quantity 25 g
Formula Weight 304.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane
EINECSNumber (5)-5567
TSCA Yes