accessibility menu, dialog, popup

UserName

CAS RN 1455-42-1

IUPAC Name: 2-[9-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]-2-methylpropan-1-ol
Synonyms: 2-[9-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]-2-methylpropan-1-ol
Molecular Weight (g/mol): 304.38
Molecular Formula: C15H28O6
InChi Key: BPZIYBJCZRUDEG-UHFFFAOYSA-N
SMILES: CC(C)(CO)C1OCC2(CO1)COC(OC2)C(C)(C)CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)