missing translation for 'onlineSavingsMsg'
Learn More

3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals

Catalog No. AAL1295814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1295814 25 g
AAL1295822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1295814 Supplier Thermo Scientific Chemicals Supplier No. L1295814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1455-42-1
Molecular Formula C15H28O6
Molecular Weight (g/mol) 304.383
MDL Number MFCD00059794
InChI Key BPZIYBJCZRUDEG-UHFFFAOYSA-N
Synonym 2,2'-2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diyl bis 2-methylpropan-1-ol, 3,9-bis 1,1-dimethyl-2-hydroxyethyl-2,4,8,10-tetraoxaspiro 5.5 undecane, cyclic acetal, 2-9-1-hydroxy-2-methylpropan-2-yl-2,4,8,10-tetraoxaspiro 5.5 undecan-3-yl-2-methylpropan-1-ol, acmc-209cve, ksc489m7h, 2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diethanol, beta3,beta3,beta9,beta9-tetramethyl, bis hydroxypivalaldehyde pentaerythritolacetal, bis hydroxypivalaldehyde pentaerythritolacetal cyclic acetal, bis hydroxypivalaldehyde pentaerythritol acetal cyclic acetal
PubChem CID 7569008
IUPAC Name 2-[3-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]-2-methylpropan-1-ol
SMILES CC(C)(CO)C1OCC2(CO1)COC(OC2)C(C)(C)CO

Specifications

Melting Point 197°C to 200°C
Quantity 25 g
Beilstein 1574570
Formula Weight 304.39
Percent Purity 97%
Chemical Name or Material 3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only